Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium chromate hydrate, 99.8% (metals basis)

Corrosion inhibitor, pigment

Manufacturer:  Alfa Aesar™ 4050322

Quantity 100g

 View more versions of this product

Catalog No. AA4050322


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16569-85-0
MFCD00801009
Magnesium chromate
140.30 (Anhydrous)
UN2923
Magnesium chromate hydrate

Specifications

16569-85-0
140.30 (Anhydrous)
UN2923
Soluble in water
Magnesium chromate hydrate
Powder
MFCD00801009
Magnesium chromate
100g
99.8%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit