Alfa Aesar™ Magnesium citrate, dibasic hydrate

Manufacturer: Alfa Aesar™ 41954A7

Quantity 5kg

 View more versions of this product

Catalog No. AA41954A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 119851-23-9
    Molecular Formula C6H6MgO7·xH2O
    Formula Weight 214.14 (Anhydrous)
    MDL Number MFCD00151706
    Merck Index 14,5663
    Chemical Name or Material Magnesium citrate dibasic hydrate

    Specifications

    Formula Weight 214.14 (Anhydrous)
    Molecular Formula C6H6MgO7·xH2O
    MDL Number MFCD00151706
    TSCA TSCA
    Physical Form Powder
    CAS 119851-23-9
    Quantity 5kg
    Merck Index 14,5663
    Color White to pale yellow
    Chemical Name or Material Magnesium citrate dibasic hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit