Alfa Aesar™ Magnesium citrate hydrate, Mg ca 16% (dry wt.)

Manufacturer: Alfa Aesar™ 41921B5

Quantity 17kg

 View more versions of this product

Catalog No. AA41921B5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3344-18-1
    Formula Weight 451.13 (Anhydrous)
    MDL Number MFCD09953657
    Merck Index 14,5663
    Chemical Name or Material Magnesium citrate hydrate

    Specifications

    Formula Weight 451.13 (Anhydrous)
    Physical Form Powder
    MDL Number MFCD09953657
    RTECS Number TZ8552900
    TSCA TSCA
    CAS 3344-18-1
    Merck Index 14,5663
    Color White to off-white
    Quantity 17kg
    Concentration Mg ≈ 16% (dry wt.)
    Chemical Name or Material Magnesium citrate hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit