Alfa Aesar™ Magnesium cube, nominally 19mm (0.75in) each side

Manufacturer: Alfa Aesar™ 45119KF

Quantity 1 piece

 View more versions of this product

Catalog No. AA45119KF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-95-4
    MDL Number MFCD00085308
    Chemical Name or Material Magnesium cube, Nominally 19mm (0.75in) each side

    Specifications

    MDL Number MFCD00085308
    CAS 7439-95-4
    Odor Odorless
    Color Silver-colored
    Weight ≈10 to 15g/cube
    Quantity 1 piece
    TSCA TSCA
    Physical Form Cubes
    Chemical Name or Material Magnesium cube, Nominally 19mm (0.75in) each side

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit