Alfa Aesar™ Magnesium ethoxide, Mg 21-22%

Manufacturer: Alfa Aesar™ 1426418

Quantity 50g

 View more versions of this product

Catalog No. AA1426418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2414-98-4
    Linear Formula Mg(OCH2CH3)2
    Formula Weight 114.44
    MDL Number MFCD00009060
    UN Number UN3205
    Beilstein 3678841
    Synonym Magnesium ethylate
    Chemical Name or Material Magnesium ethoxide

    Specifications

    Formula Weight 114.44
    Health Hazard 3 GHS P Statement
    P210-P280g-P305+P351+P338-P337+P313-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    In case of fire: Use ... for extinction.
    Store in a dry place.
    Store in a closed container.
    Physical Form Granular
    UN Number UN3205
    MDL Number MFCD00009060
    Beilstein 3678841
    Health Hazard 1 Danger
    Color Off-white
    Melting Point 270°C (dec.)
    Density 1.01g/mL
    Linear Formula Mg(OCH2CH3)2
    CAS 2414-98-4
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
    Concentration Mg 21 to 22%
    Sensitivity Air & Moisture Sensitive
    TSCA TSCA
    Synonym Magnesium ethylate
    Health Hazard 2 GHS H Statement
    H251-H319
    Self-heating: may catch fire.
    Causes serious eye irritation.
    Quantity 50g
    Odor Odorless
    Chemical Name or Material Magnesium ethoxide

    GHS H Statement
    H251-H319
    Self-heating: may catch fire.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280g-P305+P351+P338-P337+P313-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    In case of fire: Use ... for extinction.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit