Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Foil, 0.05mm (0.002in.) thick, Hard, 99.9% (Metal Basis), Alfa Aesar™

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]

Sacrificial Electrode For Other Material, Deoxidizer For Cu

Form: approx. 0.05g/25 x 25mm, Temper: as rolled
Quantity 25 x 25mm
Density 1.738g/cm3 at 20°C
Assay Percent Range 99.9%, metal basis
Odor Odorless
Physical Form Foil, 0.05mm (0.002 in.) thick, hard
Vapor Pressure 1.33 hPa (1mm Hg) at 621°C
Chemical Name or Material Magnesium

Keep container tightly closed and dry.

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : DOT Class: 4.1; Packaging Group: III

EINECSNumber : 231-104-6

TSCA : Yes

Recommended Storage : Ambient temperatures