Magnesium Foil, 0.125mm (0.005 in.) thick, hard, 99.9% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 40602FF

Quantity 25 x 25mm

 View more versions of this product

Catalog No. AA40602FF

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7439-95-4
    Physical Form Foil, 0.125mm (0.004 in.) thick
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00085308
    Chemical Name or Material Magnesium, Hard

    Specifications

    Quantity 25 x 25mm
    Color Silver
    DOT Information DOT Class: 4.1, Packaging Group: III
    EINECS Number 231-104-6
    Physical Form Foil, 0.125mm (0.004 in.) thick
    Temper Temper: as rolled
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00085308
    CAS 7439-95-4
    Odor Odorless
    Health Hazard 1 H228-H251-H261: Flammable solid. Self-heating: may catch fire. In contact with water releases flammable gas.
    Appearance Silver-colored foil
    Health Hazard 2 Keep container tightly closed and dry.
    Chemical Name or Material Magnesium, Hard

    Keep container tightly closed and dry.

    H228-H251-H261: Flammable solid. Self-heating: may catch fire. In contact with water releases flammable gas.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit