Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium foil, 1.0mm (0.04 in.) thick, hard, 99.9% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
Quantity 25 x 25mm
Weight ≈1.1g/25x25mm
UN Number UN1869
Odor Odorless
Physical Form Foil
Percent Purity 99.9%
Chemical Name or Material Magnesium foil, 1.0mm (0.04 in.) thick, Hard, Temper: as rolled
Assay (metals basis)

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

EINECSNumber : 231-104-6

TSCA : Yes

Recommended Storage : Ambient temperatures