Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium granules, -12+50 mesh, 99.8% (metals basis), Thermo Scientific™

Click to view available options
Quantity:
500g
1kg
2kg
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
Quantity 500g
Density 1.738
Assay Percent Range 99.8% (metals basis)
UN Number UN1869
Mesh Size -12+50 Mesh
Formula Weight 24.305
Physical Form Granules, ∽-12+50 Mesh
Percent Purity 99.8%
Chemical Name or Material Magnesium
Assay (metals basis)

GHS H Statement
H228-H251-H261

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

EINECSNumber : 231-104-6

RTECSNumber : OM2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only