Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hexafluorosilicate hexahydrate 96%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
18972-56-0
F6MgSi·6H2O
MFCD00150250
18972-56-0
F6MgSi·6H2O
MFCD00150250
Assay Percent Range 96%
Merck Index 15, 5732
Formula Weight 274.47
Physical Form Powder
Elemental Analysis 95% min. (ex Fluor)
Packaging Plastic bottle
Color White
Quantity 100g
Chemical Name or Material Magnesium hexafluorosilicate hexahydrate

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger