Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Acros Organics

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
7782-75-4
HMgO43H2O
174.34
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium hydrogen phosphate trihydrate,dimagnesium phosphate,unii-hf539g9l3q,magnesium phosphate dibasic trihydrate,magnesium 2+ trihydrate hydrogen phosphate,magnesium 2+ ion trihydrate hydrogen phosphate,kc defense,newberyite,magnesium dibasic phosphate trihydrate,acmc-20aklg
16204530
magnesium;hydrogen phosphate;trihydrate
O.O.O.OP(=O)([O-])[O-].[Mg+2]
7782-75-4
HMgO43H2O
174.34
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium hydrogen phosphate trihydrate,dimagnesium phosphate,unii-hf539g9l3q,magnesium phosphate dibasic trihydrate,magnesium 2+ trihydrate hydrogen phosphate,magnesium 2+ ion trihydrate hydrogen phosphate,kc defense,newberyite,magnesium dibasic phosphate trihydrate,acmc-20aklg
16204530
magnesium;hydrogen phosphate;trihydrate
O.O.O.OP(=O)([O-])[O-].[Mg+2]
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula MgHPO4·3H2O
Solubility Information Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids
Formula Weight 174.34
Percent Purity 98%
Grade Biochemistry
Chloride 0.01% max
Density 2.1300g/mL
Packaging Plastic bottle
Solubility (0.1 M in 5 M HCl) Clear colorless
Specific Gravity 2.13
Sulfate 0.03% max
Quantity 1kg
Chemical Name or Material Magnesium hydrogen phosphate trihydrate
Show More Show Less