Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium ICP Standard, 1mL = 10mg Mg (10,000ppm Mg), Ricca Chemical

Click to view available options
Quantity:
50mL
100mL
500mL
Packaging:
Poly natural
1309-48-4, 7697-37-2, 7732-18-5
MgO
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
oxomagnesium
O=[Mg]
1309-48-4, 7697-37-2, 7732-18-5
MgO
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
oxomagnesium
O=[Mg]
CAS Min % 84.0, 12.84, 1.39
CAS Max % 87.0, 13.36, 1.45
Physical Form Liquid
Packaging Poly natural
Equivalence 1 mL = 10 mg Mg
Color Colorless
Quantity 100mL
Concentration or Composition Notes 10,000ppm Mg
Chemical Name or Material Magnesium ICP Standard