Magnesium iodide, ultra dry, 99.996% (metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Manufacturer: Alfa Aesar 4431022

Quantity 100g

 View more versions of this product

Catalog No. AA4431022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10377-58-9
    Physical Form Powder
    Assay Percent Range 99.996% (Metal basis)
    Molecular Formula I2Mg
    Formula Weight 278.12
    MDL Number MFCD00049485
    Chemical Name or Material Magnesium iodide, ultra dry

    Specifications

    Physical Form Powder
    Formula Weight 278.12
    Quantity 100g
    Color White
    EINECS Number 233-825-1
    Odor Odorless
    Assay Percent Range 99.996% (Metal basis)
    Molecular Formula I2Mg
    Appearance White powder
    MDL Number MFCD00049485
    CAS 10377-58-9
    Chemical Name or Material Magnesium iodide, ultra dry

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit