Alfa Aesar™ Magnesium lactate trihydrate, 99%

Manufacturer: Alfa Aesar™ 3936936

Quantity 500g

 View more versions of this product

Catalog No. AA3936936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18917-93-6
    Formula Weight 256.49 (202.45 Anhydrous)
    MDL Number MFCD00156534
    Merck Index 14,5672
    Chemical Name or Material Magnesium lactate trihydrate

    Specifications

    Formula Weight 256.49 (202.45 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    RTECS Number OD5660000
    Merck Index 14,5672
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Color White to off-white
    Melting Point 41°C (dec.)
    TSCA TSCA
    MDL Number MFCD00156534
    CAS 18917-93-6
    Solubility Information Soluble in water, Slightly soluble in alcohol
    Health Hazard 1 Warning
    Quantity 500g
    Physical Form Powder/Lump
    Chemical Name or Material Magnesium lactate trihydrate
    Assay 99%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit