Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium methoxide, 7-8% in methanol

MgO films

Manufacturer:  Alfa Aesar™ 4044830

Quantity 250g

 View more versions of this product

Catalog No. AA4044830


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

109-88-6
MFCD00053671
Magnesium methoxide
86.38
UN1230

Specifications

7 to 8% in methanol
Alcohol-like
Colorless to pale yellow
109-88-6
Liquid
MFCD00053671
1.3380
14° (57°F)
Moisture Sensitive
250g
0.815g/mL
86.38
UN1230
Magnesium methoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H311-H331-H370-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Harmful if swallowed.

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit