Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Magnesium niobium oxide, 99.9% (metals basis)
Click to view available options
Quantity:
10g
50g
250g
Specifications
Solubility Information | Insoluble in water. |
Formula Weight | 306.11 |
Physical Form | -200 Mesh Powder |
Percent Purity | 99.9% |
Mesh Size | −200 Mesh |
Odor | Odorless |
Quantity | 10g |
Chemical Name or Material | Magnesium niobium oxide |
Assay | (metals basis) |
Safety and Handling
EINECSNumber : 235-310-7
TSCA : Yes
Recommended Storage : Ambient temperatures