Alfa Aesar™ Magnesium oxalate dihydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1299309

Quantity 10g

 View more versions of this product

Catalog No. AA1299309

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 547-66-0
    Formula Weight 148.36 (112.32 Anhydrous)
    MDL Number MFCD00287279
    UN Number UN3288
    Merck Index 14,5676
    Chemical Name or Material Magnesium oxalate dihydrate
    Grade Puratronic™

    Specifications

    Formula Weight 148.36 (112.32 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00287279
    Melting Point 150°C (dec.)
    Quantity 10g
    Solubility Information Soluble in dilute mineral acid; Practically insoluble in water; Insoluble in alcohol
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Odor Odorless
    UN Number UN3288
    Physical Form Powder
    Merck Index 14,5676
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    CAS 547-66-0
    Chemical Name or Material Magnesium oxalate dihydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit