Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxide, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
1309-48-4
MgO
40.29
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
1309-48-4
MgO
40.29
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2mg/l (20°C),reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids,insoluble in alcohol
Formula Weight 40.29
Physical Form Fine Crystalline Powder
Percent Purity 98%
Grade Pure
Density 3.5800g/mL
Packaging Plastic Bottle
Specific Gravity 3.58
Boiling Point 3600°C
Color Gray
Melting Point 2800°C
Quantity 2.5kg
Chemical Name or Material Magnesium oxide, 98%, ca 40 mesh
Show More Show Less

EINECSNumber : 215-171-9

RTECSNumber : OM3850000

TSCA : TSCA