Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Click to view available options
Quantity:
10g
50g
250g
1309-48-4
MgO
40.304
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
1309-48-4
MgO
40.304
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
Assay Percent Range 99.998% (Metal basis)
Appearance White powder
Solubility Information Soluble in acid and ammonia. Insoluble in alcohol.
Formula Weight 40.3
Physical Form -22 Mesh Powder
Density 3.58
Refractive Index 1.736
Boiling Point 3600°C
Color White
Melting Point 2852°C
Quantity 10g
Chemical Name or Material Magnesium oxide
Show More Show Less

EINECSNumber : 215-171-9

TSCA : Yes

Recommended Storage : Ambient temperatures