Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxide, ACS, 95.0% min, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
2kg
1309-48-4
MgO
40.304
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
1309-48-4
MgO
40.304
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
Assay Percent Range 95.0% min.
Appearance White granules
Solubility Information Soluble in acid and ammonia. Insoluble in alcohol.
Formula Weight 40.3
Physical Form Granules
Grade ACS
Density 3.58 g/cm3 at 20°C
Refractive Index 1.736
Boiling Point 3600°C
Color White
Melting Point 2852°C
Quantity 100g
Chemical Name or Material Magnesium oxide
Show More Show Less

EINECSNumber : 215-171-9

TSCA : Yes

Recommended Storage : Ambient temperatures