Learn More
Magnesium oxide, ACS, 95.0% min, Thermo Scientific Chemicals
Description
Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 3600°C |
Melting Point | 2852°C |
Color | White |
Physical Form | Granules |
Quantity | 100 g |
Assay Percent Range | 95.0% min. |
Solubility Information | Soluble in acid and ammonia. Insoluble in alcohol. |
Formula Weight | 40.3 |
Grade | ACS |
Refractive Index | 1.736 |
Show More |
Safety and Handling
EINECSNumber | 215-171-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.