Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxide, Powder, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2kg
500g
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

1309-48-4
MgO
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
CAS 1309-48-4
Molecular Formula MgO
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES O=[Mg]
IUPAC Name oxomagnesium
Molecular Weight (g/mol) 40.304
PubChem CID 14792
Formula Weight 40.3
Physical Form Powder
Packaging Poly Pail
Specific Gravity 1L = 3.58kg
Quantity 2kg
Chemical Name or Material Magnesium Oxide
Grade FCC/USP
Show More Show Less