Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Magnesium oxide, purum p.a., Honeywell

Click to view available options
Quantity:
250g
1kg
1309-48-4
MgO
40.304
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
1309-48-4
MgO
40.304
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]

Purum p.a., heavy,≥98.0% (based on calcined substance, KT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Linear Formula MgO
UN Number NONH for all modes of transport
Formula Weight 40.3g/mol
Percent Purity ≥98% (based on calcined substance, KT)
Grade purum p.a.
Quantity 1kg
Chemical Name or Material Magnesium oxide

RTECSNumber : OM3850000