Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis excluding Ca)
Click to view available options
Quantity:
1 ea.
Specifications
MDL Number | MFCD00011109 |
Formula Weight | 40.3 |
Percent Purity | 99.95% |
Quantity | 1 ea. |
Chemical Name or Material | Magnesium oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick |
Assay | (metals basis excluding Ca) |
Safety and Handling
EINECSNumber : 215-171-9
TSCA : Yes
Recommended Storage : Ambient temperatures