Alfa Aesar™ Magnesium Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Alfa Aesar™

Manufacturer: Alfa Aesar™ 41436KU

Quantity 5 ea.

 View more versions of this product

Catalog No. AA41436KU

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Molecular Formula MgO
    MDL Number MFCD00011109
    Chemical Name or Material Magnesium Oxide Substrate, Polished Two Sides

    Specifications

    Physical Form Plate
    Color White
    Quantity 5 ea.
    Odor Odorless
    Molecular Formula MgO
    MDL Number MFCD00011109
    Chemical Name or Material Magnesium Oxide Substrate, Polished Two Sides

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit