Alfa Aesar™ Magnesium ribbon, approximate dimensions, 3.0mm (0.12in) wide, 0.15mm (0.0059 in.) thick

Manufacturer: Alfa Aesar™ 1281151

Quantity 5 x 25g

 View more versions of this product

Catalog No. AA1281151

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-95-4
    MDL Number MFCD00085308
    UN Number UN1869
    Chemical Name or Material Magnesium ribbon, Approximate dimensions: 3.0mm (0.12 in.) wide, 0.15mm (0.0059 in.) thick

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P231+P232-P280-P370+P378i-P420-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store away from other materials.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H228-H251-H261
    Flammable solid.
    Self-heating: may catch fire.
    In contact with water releases flammable gas.
    TSCA TSCA
    MDL Number MFCD00085308
    CAS 7439-95-4
    Odor Odorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM
    UN Number UN1869
    Weight ≈0.78g/m
    Quantity 5 x 25g
    Chemical Name or Material Magnesium ribbon, Approximate dimensions: 3.0mm (0.12 in.) wide, 0.15mm (0.0059 in.) thick

    GHS H Statement
    H228-H251-H261
    Flammable solid.
    Self-heating: may catch fire.
    In contact with water releases flammable gas.

    GHS P Statement
    P210-P231+P232-P280-P370+P378i-P420-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store away from other materials.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit