Alfa Aesar™ Magnesium salicylate tetrahydrate, tech.

Manufacturer: Alfa Aesar™ 39379A4

Quantity 2.5kg

 View more versions of this product

Catalog No. AA39379A4

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18917-95-8
    Formula Weight 370.60 (298.60 Anhydrous)
    MDL Number MFCD00045815
    Merck Index 14,5672
    Chemical Name or Material Magnesium salicylate tetrahydrate
    Grade Technical Grade

    Specifications

    Formula Weight 370.60 (298.60 Anhydrous)
    MDL Number MFCD00045815
    Odor Odorless
    Physical Form Powder
    CAS 18917-95-8
    Merck Index 14,5672
    Solubility Information Soluble in water and alcohol
    Sensitivity Air Sensitive
    Quantity 2.5kg
    Chemical Name or Material Magnesium salicylate tetrahydrate
    Grade Technical Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit