Magnesium silicate monohydrate (Talc), Alfa Aesar™

Lubricant, Filler, Electric and Heat Insulator

Manufacturer: Alfa Aesar 4031836

Quantity 500g

 View more versions of this product

Catalog No. AA4031836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14807-96-6
    Physical Form Powder
    Molecular Formula 3MgO·4SiO2·H2O
    Formula Weight 379.28 (361.27 Anhydrous)
    MDL Number MFCD00792903
    Chemical Name or Material Magnesium silicate monohydrate (Talc)

    Specifications

    Physical Form Powder
    Formula Weight 379.28 (361.27 Anhydrous)
    Quantity 500g
    Color White to pale gray
    EINECS Number 238-877-9
    Odor Odorless
    Molecular Formula 3MgO·4SiO2·H2O
    Appearance White to pale gray powder
    MDL Number MFCD00792903
    CAS 14807-96-6
    Health Hazard 1 H332: Harmful if inhaled.
    Chemical Name or Material Magnesium silicate monohydrate (Talc)

    H332: Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit