Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate heptahydrate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
25kg
Packaging:
Fibre drum
Plastic bottle
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Assay Percent Range 99.4% min. (Complexometry)
Linear Formula MgSO4·7H2O
Merck Index 15, 5755
Solubility Information Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol,sparingly soluble in alcohol
Formula Weight 246.48
Physical Form Crystals
Percent Purity 99.5%
Grade Analysis
Cadmium (Cd) 5ppm max.
Calcium (Ca) 50ppm max.
Chloride 5ppm max
Chlorine (Cl) 5ppm max.
Cobalt (Co) 5ppm max.
Heavy Metals (as Pb) 5ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iron (Fe) 5ppm max.
Manganese (Mn) 5ppm max.
Packaging Fibre drum
Potassium (K) 50ppm max.
Sodium (Na) 200ppm max.
Solubility Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
Trace Analysis 50ppm max.
Color White
Melting Point 770.0°C
pH 5 to 8 (5% soln. at 25°C)
Quantity 25kg
Chemical Name or Material Magnesium sulfate heptahydrate, p.a.
Show More Show Less

RTECSNumber : OM4508000