Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate, Reagent grade, Honeywell

Click to view available options
Quantity:
500g
2kg
2.5kg
7487-88-9
MgO4S
120.361
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate,magnesium sulphate,magnesium sulfate anhydrous,sulfuric acid magnesium salt 1:1,bitter salt,sal angalis,magnesium sulfate 1:1,mgso4,sal de sedlitz,tomix ot
24083
CHEBI:32599
magnesium;sulfate
[O-]S(=O)(=O)[O-].[Mg+2]
7487-88-9
MgO4S
120.361
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate,magnesium sulphate,magnesium sulfate anhydrous,sulfuric acid magnesium salt 1:1,bitter salt,sal angalis,magnesium sulfate 1:1,mgso4,sal de sedlitz,tomix ot
24083
CHEBI:32599
magnesium;sulfate
[O-]S(=O)(=O)[O-].[Mg+2]

Reagent Grade, anhydrous,≥97%

  • ≥95% for general laboratory use.
UN Number NONH for all modes of transport
Formula Weight 120.37g/mol
Percent Purity ≥97%
Grade Reagent
Vapor Pressure 0.1mmHg (20°C)
Vapor Density <0.01 (vs air)
Physical State Anhydrous
Quantity 2kg
Chemical Name or Material Magnesium Sulfate

EINECSNumber : 231-298-2

RTECSNumber : OM4500000