Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate Solution EP, Ricca Chemical

Click to view available options
Quantity:
500mL
7732-18-5,10034-99-8,7664-93-9
73.0,18.14,7.34
75.0,18.88,7.64
MAGNESIUM SULFATE SOLUTION
7732-18-5,10034-99-8,7664-93-9
73.0,18.14,7.34
75.0,18.88,7.64
MAGNESIUM SULFATE SOLUTION
Synonym dihydrogen oxide,dihydrogen monoxide
Grade EP
Quantity 500mL