Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium tert-butoxide, 93+%, Acros Organics

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass Bottle
Plastic bottle
32149-57-8
C8H18MgO2
170.53
MFCD03788761
RTKCPZYOLXPARI-UHFFFAOYSA-N
magnesium tert-butoxide,magnesium di-tert-butoxide,magnesium 2-methylpropan-2-olate,magnesium t-butoxide,magnesiumtert-butoxide,di-tert-butoxymagnesium,magnesium tert. butoxide,ksc226k7n,magnesium 2+ bis 2-methylpropan-2-olate
12040442
magnesium;2-methylpropan-2-olate
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]
32149-57-8
C8H18MgO2
170.53
MFCD03788761
RTKCPZYOLXPARI-UHFFFAOYSA-N
magnesium tert-butoxide,magnesium di-tert-butoxide,magnesium 2-methylpropan-2-olate,magnesium t-butoxide,magnesiumtert-butoxide,di-tert-butoxymagnesium,magnesium tert. butoxide,ksc226k7n,magnesium 2+ bis 2-methylpropan-2-olate
12040442
magnesium;2-methylpropan-2-olate
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]
Packaging Glass Bottle
Color White to Gray
Quantity 10g
Assay Percent Range 93+%
Solubility Information Solubility in water: reacts violently
Formula Weight 170.53
Physical Form Powder
Percent Purity 93+%
Chemical Name or Material Magnesium tert-butoxide

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

EINECSNumber : 250-931-3