Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium tert-butoxide, 93+%, Thermo Scientific Chemicals

Catalog Number AC390470500
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
32149-57-8
C8H18MgO2
170.53
MFCD03788761
RTKCPZYOLXPARI-UHFFFAOYSA-N
magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
12040442
magnesium;2-methylpropan-2-olate
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]
32149-57-8
C8H18MgO2
170.53
MFCD03788761
RTKCPZYOLXPARI-UHFFFAOYSA-N
magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
12040442
magnesium;2-methylpropan-2-olate
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Gray
Assay Percent Range 93+%
Packaging Plastic bottle
Quantity 50 g
Solubility Information Solubility in water: reacts violently
Formula Weight 170.53
Percent Purity 93+%
Physical Form Powder
Chemical Name or Material Magnesium tert-butoxide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 250-931-3

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit