Alfa Aesar™ Magnesium titanium oxide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 1139830

Quantity 250g

 View more versions of this product

Catalog No. AA1139830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12032-30-3
    Formula Weight 120.21
    MDL Number MFCD00016198
    Synonym Magnesium titanate
    Chemical Name or Material Magnesium titanium oxide

    Specifications

    Formula Weight 120.21
    MDL Number MFCD00016198
    Odor Odorless
    Particle Size 2 Micron
    Synonym Magnesium titanate
    Color White
    Melting Point 1610°C
    TSCA TSCA
    Physical Form Powder
    CAS 12032-30-3
    Quantity 250g
    Density 3.36g/mL
    Chemical Name or Material Magnesium titanium oxide
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit