Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium turnings, 1cm (0.4 in.) & down, Puratronic™, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 1063014

Quantity 25g

 View more versions of this product

Catalog No. AA1063014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7439-95-4
UN1869
Puratronic™
MFCD00085308
Magnesium turnings

Specifications

25g
MFCD00085308
Odorless
Magnesium turnings
Puratronic™
7439-95-4
UN1869
1cm (0.4 in.) & down
99.98%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

GHS P Statement
P210-P231+P232-P280-P370+P378i-P420-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store away from other materials.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit