Learn More
Cole-Parmer O C White 52400-4 Large-diameter Circular Illuminated Magnifier, 2x magnification  DFS Item
Manufacturer:  Cole-Parmer 4180785
Large 7-1/2" diameter lens and full-spectrum fluorescent bulb provide excellent viewing. Heavy-duty construction includes 1/2" square arms with smooth spring-loaded movement, cast-iron clamp, and ESD-safe shade. The 10" (25.4 cm) dia. circular, 34-watt bulb provides shadowless illumination. Swivel mount allows sideways and angular mounting for nearly limitless positioning. Magnifiers operate on 120 VAC.
Catalog No. 50-631-830