Learn More
Carson™ MagniLamp Pro Desk Lamp Magnifier Products for Science Education
MagniLamp Pro Llighted Desk Lamp Magnifier is an adjustable magnifying lamp.
Manufacturer:  Carson™ CP80
Catalog No. S81170
Add to cart
Description
Description
  • Contains a 5-in. (130 mm) glass lens which allows for very high light transmission and provides an extremely sharp and clear image.
  • Glass lenses are difficult to scratch, and magnify slightly more than acrylic magnifiers as a result of the material density.
  • Fully adjustable head that extends to 10 in.
  • Use the built-in handle on the magnifier head to rotate the lens to meet any angle for viewing needs.
  • 48 super bright, built-in LED lights
  • Plug into a standard outlet using the included AC/DC adapter
  • Comes with a protective cover to eliminate scratches and dust when not in-use.
  • Ideal for professional or personal use - for desk, workbench, table or hobby station.
  • Ideal inspection magnifier for multiple tasks such as model building, arts and crafts, jewelry design, soldering, small electronics, coin and stamp collectors, sewing, knitting, reading books, newspapers, magazines, coupons, labels and other low vision needs.
Specifications
Specifications
Magnifiers
LED
2.5X