Learn More
NVIGEN INC MAGVIGEN-ANTI-FLAG MOUSE 1 M  DFS Item
Manufacturer:  NVIGEN INC 51007
Beads, MagVigen-anti-Flag, mouse magnetic nanoparticles, High binding capcity and specificity. Applications: Immunoprecipitation, cell isolation, antibody/protein purification.
Catalog No. 50-672-447