Learn More
Sklar InstrumentsSupplier Diversity Partner Maier Gross Uterine Dressing Forceps, Sklar, 10 in., Curved, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 908142
Maier Gross Uterine Dressing Forceps, Sklar, 10 in., Curved, Serrated, Stainless Steel. Sklar's Maier-Gross Dressing Forceps are commonly used for holding, placement, and removal of dressings. Available in multiple lengths and curvatures, serrated tips provide a secure grip on swabs, gauze, cotton, and other dressing materials. These forceps are also available with or without ratcheted handles. This product is curved with fenestrated, serrated tips, a non-ratcheted handle, and a length of 10 inches.
Catalog No. 50-143-6858