Learn More
3M™ Versaflo™ S-Series Hood with Faceseal, Integrated Head Suspension
Versaflo™ S-Series Hood with integrated suspension provides head, face, neck, and shoulder coverage as well as providing eye and face protection from liquid splash and low energy flying particles and dust.
Specifications
Certifications/Compliance | NIOSH |
---|---|
Color | White |
For Use With (Application) | Assembly, facility maintenance, machine operation, painting, sanding |
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
19-805-614
|
3M™
7000029981 |
S-433 L-5 |
Case of 5 for $508.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
19-805-600
|
3M™
7000127432 |
S-533 S |
Case for $124.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
19-805-599
|
3M™
7000052767 |
S-533 L |
Case for $124.00
|
N/A
![]() |
|||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
19-805-615
|
3M™
7000127431 |
S-433S-5 |
Case of 5 for $388.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Integrated head suspension provides head, face, neck and shoulder coverage.
- Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F).
- Suitable for applications which require frequent replacement of the entire headtop assembly or those where there is less soiling of the outer fabric.
- No assembly required
- Ready to use straight out of the box.
Specifications
NIOSH | |
White | |
Assembly, facility maintenance, machine operation, painting, sanding |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.