Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141004
This item is not returnable.
View return policy
Description
MAL Polyclonal Antibody for Western Blot, IHC (P)
MAL, a nonglycosylated integral membrane proteolipid, has been identified as a major internal component of glycolipid-enriched membrane (GEM) microdomains. MAL is the first integral membrane proteolipid component to have been shown as a necessary element in the cellular apical sorting apparatus for newly synthesized proteins. In its steady state this highly hydrophobic proteolipid, resides predominantly in the apical zone of polarized epithelial cells. It has also been shown in itinerant protein cycling between the plasma membrane and endosomes and in other cells. This 17-kDa proteolipid contains four putative transmembrane domains. These domains along with its protein trafficking properties are defined by the specific signals delegated by its specific amino acid sequence.
Specifications
MAL | |
Polyclonal | |
Unconjugated | |
MAL | |
17 kDa myelin vesicular protein; Mal; mal T-cell differentiation protein; mal, T cell differentiation protein L homeolog; mal, T-cell differentiation protein; mal.L; MALGENE; Mpv17; Mvp17; MyD88-adapter-like; myelin and lymphocyte protein; myelin and lymphocyte protein, T cell differentiation protein; myelin and lymphocyte protein, T-cell differentiation protein; NS 3; T-lymphocyte maturation-associated protein; Vip17; XELAEV_18026637mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17153, 25263, 4118 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O09198, P21145, Q64349 | |
MAL | |
Synthetic peptide within human MAL aa 103-146. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction