Available on GSA/VA Contract for Federal Government customers only. Malachite Green Oxalate (Certified Biological Stain), Fisher Chemical

$120.48

Chemical Identifiers

Chemical Name or Material Malachite Green Oxalate
CAS [547-58-0]
Purity Grade Certified Biological Stain
Molecular Formula C14H14N3O3SNa
Formula Weight 327.33
MFCD Number MFCD00011766
UN Number 2811
Synonym Basic Green 4
View More Specs

  • Products 1
  • Description & Specifications
  • Safety and Handling
Products
Catalog Number Description Price Quantity Availability  

M-29025

Available on GSA/VA Contract for Federal Government customers only.  SDS
Glass Bottle; 25g
Each for $120.48
Case of 6 EA for $542.15
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products