Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malaria pLDH Mouse anti-Protozoa, Clone: HM088, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13613

Catalog No. 89-033-576


Add to cart

Description

Description

Mouse monoclonal antibody raised against malaria pLDH.

  • High specificity
  • High reactivity
Specifications

Specifications

Malaria pLDH
HM088
Mouse monoclonal antibody raised against malaria pLDH.
In 10 mM PBS, pH 7.4.
Mouse
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Lateral Flow
Unconjugated
Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Recombinant malaria pLDH antigen.
RUO
Primary
Protozoa
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)