Thermo Scientific™ Male Luer-LOK Priming Syringes Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Manufacturer: Thermo Scientific™ 365LL541
 View more versions of this product
Catalog No. 03-170-301
  Check Availability
Add to Cart
Alert
f
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Volume (Metric) 2.5mL
    Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
    • Square tip to prevent damage to the injector
    • Wide range of volumes
    • Precision made from borosilicate glass and stainless steel
    • Robust design and easy-to-read markings
  • Description & Specifications
Related Products
Product Content Feedback