Learn More
Microsolv Technology CorporationSupplier Diversity Partner MALE NUTS SS SHORT 10/PK DFS Item
Manufacturer:  Microsolv Technology CorporationSupplier Diversity Partner 4959001
Male Nuts, HPLC, 316L stainless steel. 10-32 hex head for use on short Rheodyne threads. 10/pack.
Catalog No. 50-014-51725