Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Male Rotating Luer Lock and Nut Assembly with 5/32 in. ID - Medical Nylon - QC

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked and rotate as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 

$30.00

Specifications

Material Natural Nylon
Type Rotating Luer Lock
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Description Class 7 Cleanroom Manufactured
Certifications/Compliance Compliance Certificates Available
View More Specs

Products 7
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
01-000-263
View Documents
Natural
Pack of 100 for $30.00
Add to cart
 
01-000-261
View Documents
Berry Blue
Pack of 100 for $30.00
Add to cart
 
01-000-262
View Documents
Lime Green
Pack of 100 for $30.00
Add to cart
 
01-000-264
View Documents
Orange
Pack of 100 for $30.00
Add to cart
 
01-000-265
View Documents
Rose Red
Pack of 100 for $30.00
Add to cart
 
01-000-266
View Documents
Violet Purple
Pack of 100 for $30.00
Add to cart
 
01-000-267
View Documents
Yellow
Pack of 100 for $30.00
Add to cart
 
Description

Description

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Specifications

Natural Nylon
Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Compliance Certificates Available
Rotating Luer Lock
Class 7 Cleanroom Manufactured
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit