Learn More
Fisherbrand™ Male Stationary Luer Lock and Nut Assembly with 3/32 in. ID - Medical Nylon - QC

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant.
$60.50
Specifications
Material | Natural Nylon |
---|---|
Type | Stationary Luer Lock |
For Use With (Application) | Tubing connections; Medical device input/output ports; Drug and fluid transfer lines |
Description | Class 7 Cleanroom Manufactured |
Certifications/Compliance | Compliance Certificates Available |
Catalog Number | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Color | Price | Quantity & Availability | ||||||
01-000-366
![]() |
Berry Blue |
Pack of 100 for $60.50
|
|
01-000-369
![]() |
Orange |
Pack of 100 for $60.50
|
|
01-000-367
![]() |
Lime Green |
Pack of 100 for $60.50
|
|
01-000-368
![]() |
Natural |
Pack of 100 for $60.50
|
|
01-000-370
![]() |
Rose Red |
Pack of 100 for $60.50
|
|
01-000-371
![]() |
Violet Purple |
Pack of 100 for $60.50
|
|
01-000-372
![]() |
Yellow |
Pack of 100 for $60.50
|
|
Description
- Single & Multi Use Applications
- PVC-Free
- ISO 80369-7 Compliant
- Superior Single Barb Design
- USP Class VI Compliant
- USP 661, USP81, USP 88
- RoHS Compliant
- Gamma Stable
- Animal Derivative Free
- Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Natural Nylon | |
Tubing connections; Medical device input/output ports; Drug and fluid transfer lines | |
Compliance Certificates Available |
Stationary Luer Lock | |
Class 7 Cleanroom Manufactured |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.