Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Maleate, 0.2M buffer soln., pH 6.0

Click to view available options
Quantity:
250mL
500mL
3105-55-3
C4H9NaO7
192.099
MFCD00012466
VOWLSUZYJXDGNJ-SQDRRJMKSA-M
91658932
sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate
C(=CC(=O)[O-])C(=O)O.O.O.O.[Na+]
3105-55-3
C4H9NaO7
192.099
MFCD00012466
VOWLSUZYJXDGNJ-SQDRRJMKSA-M
91658932
sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate
C(=CC(=O)[O-])C(=O)O.O.O.O.[Na+]
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 6.0
Color Colorless
Quantity 500mL
Chemical Name or Material Maleate buffer soln.