Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic Acid, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
110-16-7
C4H4O4
116.072
MFCD00063177
VZCYOOQTPOCHFL-UPHRSURJSA-N
maleic acid,cis-butenedioic acid,toxilic acid,maleinic acid,malenic acid,2z-but-2-enedioic acid,2-butenedioic acid z,maleate,cis-1,2-ethylenedicarboxylic acid,z-butenedioic acid
444266
CHEBI:18300
(Z)-but-2-enedioic acid
C(=CC(=O)O)C(=O)O
110-16-7
C4H4O4
116.072
MFCD00063177
VZCYOOQTPOCHFL-UPHRSURJSA-N
maleic acid,cis-butenedioic acid,toxilic acid,maleinic acid,malenic acid,2z-but-2-enedioic acid,2-butenedioic acid z,maleate,cis-1,2-ethylenedicarboxylic acid,z-butenedioic acid
444266
CHEBI:18300
(Z)-but-2-enedioic acid
C(=CC(=O)O)C(=O)O
Assay Percent Range 99%
Linear Formula HO2CCH=CHCO2H
Beilstein 02, IV, 2199
Merck Index 15, 5767
Formula Weight 116.07
Physical Form Crystalline Powder
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: 790g/L (25°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol and acetone, slightly soluble in ether and benzene, practically insoluble in carbon tetrachloride
Water 2% max.
Melting Point 134.0°C to 138.0°C
Color White
Quantity 1kg
Flash Point 127°C
Chemical Name or Material Maleic acid
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.