Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic Acid, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Poly Bottle
110-16-7
C4H4O4
116.072
VZCYOOQTPOCHFL-UPHRSURJSA-N
maleic acid,cis-butenedioic acid,toxilic acid,maleinic acid,malenic acid,2z-but-2-enedioic acid,2-butenedioic acid z,maleate,cis-1,2-ethylenedicarboxylic acid,z-butenedioic acid
444266
CHEBI:18300
(Z)-but-2-enedioic acid
C(=CC(=O)O)C(=O)O
110-16-7
C4H4O4
116.072
VZCYOOQTPOCHFL-UPHRSURJSA-N
maleic acid,cis-butenedioic acid,toxilic acid,maleinic acid,malenic acid,2z-but-2-enedioic acid,2-butenedioic acid z,maleate,cis-1,2-ethylenedicarboxylic acid,z-butenedioic acid
444266
CHEBI:18300
(Z)-but-2-enedioic acid
C(=CC(=O)O)C(=O)O

A grade of chemicals for laboratory use

Formula Weight 116.07
Packaging Poly Bottle
Specific Gravity 1L = 1.59kg
Quantity 1kg
Chemical Name or Material Maleic Acid